missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyl Phthalate 98.0+%, TCI America™

Catalog No. P030425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P030425ML 25 mL
P0304500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P030425ML Supplier TCI America Supplier No. P030425ML

Chemical Identifiers

CAS 117-84-0
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.564
MDL Number MFCD00015292
InChI Key MQIUGAXCHLFZKX-UHFFFAOYSA-N
Synonym dioctyl phthalate, di-n-octyl phthalate, dinopol nop, n-octyl phthalate, vinicizer 85, phthalic acid, dioctyl ester, polycizer 162, dnop, phthalic acid di-n-octyl ester, dioctyl 1,2-benzenedicarboxylate
PubChem CID 8346
ChEBI CHEBI:34679
IUPAC Name dioctyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCC

Specifications

Color Yellow
Boiling Point 380°C
Quantity 25 mL
Formula Weight 390.56
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Di-n-octyl Phthalate
EINECSNumber (3)-1307
RTECSNumber TI1925000
TSCA Yes