missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyl phthalate, 98%

Catalog No. AA41522AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41522AC 25 mL
AA41522AE 100 mL
AA41522AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41522AC Supplier Thermo Scientific Chemicals Supplier No. 041522AC

Plasticizer | CAS: 117-84-0 | C24H38O4 | 390.564 g/mol

Di-n-octyl phthalate is used as a plasticizer for many resins and elastomers. It acts as an additive. It is also used for medical tubing and blood storage bags, wire and cables, carpet back coating, floor tile and in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 117-84-0
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.564
MDL Number MFCD00015292
InChI Key MQIUGAXCHLFZKX-UHFFFAOYSA-N
Synonym dioctyl phthalate, di-n-octyl phthalate, dinopol nop, n-octyl phthalate, vinicizer 85, phthalic acid, dioctyl ester, polycizer 162, dnop, phthalic acid di-n-octyl ester, dioctyl 1,2-benzenedicarboxylate
PubChem CID 8346
ChEBI CHEBI:34679
IUPAC Name dioctyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCC

Specifications

Density 0.98g/mL
Assay Percent Range 98%
Quantity 25 mL
Merck Index 14,2864
Formula Weight 390.56
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Di-n-octyl phthalate
RTECSNumber TI1925000
TSCA TSCA

RUO – Research Use Only