missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyl phenylphosphonate, 97%

Catalog No. p-7021220
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1346806 5 g
AAA1346814 25 g
AAA1346822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1346806 Supplier Thermo Scientific Chemicals Supplier No. A1346806
Only null left

CAS: 1754-47-8 | C22H39O3P | 382.53 g/mol

Di-n-octyl phenylphosphonate acts as a plasticizer, which is useful in ion-selective electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-octyl phenylphosphonate acts as a plasticizer, which is useful in ion-selective electrodes.

Solubility
Miscible with tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1754-47-8
Molecular Formula C22H39O3P
Molecular Weight (g/mol) 382.53
MDL Number MFCD00009561
InChI Key HAKMAMKAFTZXOZ-UHFFFAOYSA-N
Synonym dioctyl phenylphosphonate, di-n-octyl phenylphosphonate, dioctylphenylphosphonate, phenylphosphonic acid dioctyl ester, benzenephosphonic acid, dioctyl ester, di-n-octyl phenyl phosphonate, dioctyl phenyl phosphonate, phosphonic acid, phenyl-, dioctyl ester, phosphonic acid, p-phenyl-, dioctyl ester, di-n-octylphenylphosphonat
PubChem CID 15641
IUPAC Name dioctoxyphosphorylbenzene
SMILES CCCCCCCCOP(=O)(OCCCCCCCC)C1=CC=CC=C1

Specifications

Density 0.932
Boiling Point 219°C to 220°C (0.7 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4795
Quantity 5 g
Beilstein 2948496
Solubility Information Miscible with tetrahydrofuran.
Formula Weight 382.53
Percent Purity 97%
Chemical Name or Material Di-n-octyl phenylphosphonate
EINECSNumber 217-142-6
RTECSNumber TA0379000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only