Learn More
Di-n-octadecyl disulfide, 98%
CAS: 2500-88-1 | C36H74S2 | 571.11 g/mol
Supplier: Thermo Scientific Chemicals B2197806
Description
It is used as a co stabilizer, antioxidant for plastics, polyoletins, HDPE, LDPE, PMMA, PA, engineering thermoplastic elastomers, synergist, thermal stabilizer, processing stabilizer in high temperature applications, peroxide decomposer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a co stabilizer, antioxidant for plastics, polyoletins, HDPE, LDPE, PMMA, PA, engineering thermoplastic elastomers, synergist, thermal stabilizer, processing stabilizer in high temperature applications, peroxide decomposer.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications
| Di-n-octadecyl disulfide | |
| 60°C to 64°C | |
| C36H74S2 | |
| 5 g | |
| disulfide, dioctadecyl, dioctadecyl disulfide, octadecyl disulfide, di-n-octadecyl disulfide, distearyl disulfide, 1-octadecyldisulfanyl octadecane, dioctadecyl disulphide, di-n-octadecyldisulfide, dioctadecyldisulfid, n-octadecyl disulfide | |
| MQQKTNDBASEZSD-UHFFFAOYSA-N | |
| 1-(octadecyldisulfanyl)octadecane | |
| 75627 | |
| 98% |
| 2500-88-1 | |
| Unpleasant | |
| MFCD00026687 | |
| 1715376 | |
| Insoluble in water. | |
| CCCCCCCCCCCCCCCCCCSSCCCCCCCCCCCCCCCCCC | |
| 571.11 | |
| 571.11 |
Chemical Identifiers
| 2500-88-1 | |
| 571.11 | |
| MQQKTNDBASEZSD-UHFFFAOYSA-N | |
| 75627 | |
| CCCCCCCCCCCCCCCCCCSSCCCCCCCCCCCCCCCCCC |
| C36H74S2 | |
| MFCD00026687 | |
| disulfide, dioctadecyl, dioctadecyl disulfide, octadecyl disulfide, di-n-octadecyl disulfide, distearyl disulfide, 1-octadecyldisulfanyl octadecane, dioctadecyl disulphide, di-n-octadecyldisulfide, dioctadecyldisulfid, n-octadecyl disulfide | |
| 1-(octadecyldisulfanyl)octadecane |
Safety and Handling
EINECSNumber : 219-702-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only