missing translation for 'onlineSavingsMsg'
Learn More

Di-n-hexyl ether, 98%

Catalog No. AAL0707609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0707609 10 g
AAL0707618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0707609 Supplier Thermo Scientific Chemicals Supplier No. L0707609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112-58-3 | C12H26O | 186.34 g/mol

Dihexyl ether has been used as: supported liquid membrane during hollow-fiber liquid-phase microextraction method for determination of nitrophenolic compounds from atmospheric aerosol particles, an extraction solvent to detect avermectins in stream water by hollow-fiber-assisted liquid-phase microextraction technique coupled with LC-MS/MS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dihexyl ether has been used as: supported liquid membrane during hollow-fiber liquid-phase microextraction method for determination of nitrophenolic compounds from atmospheric aerosol particles, an extraction solvent to detect avermectins in stream water by hollow-fiber-assisted liquid-phase microextraction technique coupled with LC-MS/MS.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.

Chemical Identifiers

CAS 112-58-3
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
MDL Number MFCD00009525
InChI Key BPIUIOXAFBGMNB-UHFFFAOYSA-N
Synonym dihexyl ether, hexyl ether, hexane, 1,1'-oxybis, n-hexyl ether, di-n-hexyl ether, ether, dihexyl, bis 1-hexyl ether, 1,1'-oxybis hexane, 1-hexyloxy hexane
PubChem CID 8198
SMILES CCCCCCOCCCCCC

Specifications

Melting Point -43°C
Density 0.793
Boiling Point 228°C to 230°C
Flash Point 78°C (172°F)
Odor Mild
Refractive Index 1.4204
Linear Formula CH3(CH2)5O(CH2)5CH3
Quantity 10 g
Beilstein 1738177
Solubility Information Fully miscible in water.
IUPAC Name 1-hexoxyhexane
Formula Weight 186.34
Percent Purity 98%
Chemical Name or Material Di-n-hexyl ether
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 203-987-8
RTECSNumber KN8600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only