missing translation for 'onlineSavingsMsg'
Learn More

Di-n-decyl sulfide, 97%

Catalog No. p-7026527
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2256009 10 g
AAB2256018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2256009 Supplier Thermo Scientific Chemicals Supplier No. B2256009
Only null left

CAS: 693-83-4 | C20H42S | 314.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 693-83-4
Molecular Formula C20H42S
Molecular Weight (g/mol) 314.62
MDL Number MFCD00026542
InChI Key RKYMVQJWYYOIJB-UHFFFAOYSA-N
Synonym decyl sulfide, didecyl sulfide, di-n-decyl sulfide, decane, 1,1'-thiobis, n-decyl sulfide, 11-thiaheneicosane, decylthiodecane, didecyl sulphide, di-n-decylsulfide, decane,1'-thiobis
PubChem CID 69662
IUPAC Name 1-decylsulfanyldecane
SMILES CCCCCCCCCCSCCCCCCCCCC

Specifications

Melting Point 25°C to 28°C
Boiling Point 217°C to 218°C (8 mmHg)
Quantity 10 g
Beilstein 1763427
Formula Weight 314.63
Percent Purity 97%
Chemical Name or Material Di-n-decyl sulfide
EINECSNumber 211-761-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only