missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyl sulfoxide, 97%

Catalog No. p-7027144
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2367206 5 g
AAB2367214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2367206 Supplier Thermo Scientific Chemicals Supplier No. B2367206
Only null left

CAS: 2168-93-6 | C8H18OS | 162.291 g/mol

It is used as an active pharmaceutical intermediate and in the extraction of thorium(IV) from nitric acid solutions in xylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate and in the extraction of thorium(IV) from nitric acid solutions in xylene.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.

Chemical Identifiers

CAS 2168-93-6
Molecular Formula C8H18OS
Molecular Weight (g/mol) 162.291
MDL Number MFCD00002093
InChI Key LOWMYOWHQMKBTM-UHFFFAOYSA-N
Synonym dibutyl sulfoxide, butyl sulfoxide, n-butyl sulfoxide, dibutyl sulphoxide, di-n-butyl sulfoxide, butane, 1,1'-sulfinylbis, sulfoxide, dibutyl, di-n-butylsulfoxide, butane,1-butylsulfinyl, butylsulfinyl butane
PubChem CID 16568
IUPAC Name 1-butylsulfinylbutane
SMILES CCCCS(=O)CCCC

Specifications

Melting Point 31°C to 34°C
Density 0.832
Boiling Point 250°C
Flash Point 121°C (249°F)
Refractive Index 1.467
Quantity 5 g
Beilstein 1747425
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 162.3
Percent Purity 97%
Chemical Name or Material Di-n-butyl sulfoxide
Show More Show Less
EINECSNumber 218-511-4
RTECSNumber EK5700000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only