missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyl phthalate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A132570C

 View more versions of this product

Catalog No. AAA132570C

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a general purpose monomeric phthalate with excellent solubilizing characteristics. It is commonly used as an additive to adhesives and printing inks. It also finds utility in shower curtains, raincoats and vinyl coated fabrics.

Solubility
Soluble in water. (0.4 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications

Specifications

Di-n-butyl phthalate
-35°C
339°C to 340°C
Odorless
1.492
10,000 g
1914064
dibutyl phthalate, di-n-butyl phthalate, n-butyl phthalate, butyl phthalate, celluflex dpb, elaol, genoplast b, palatinol c, polycizer dbp, unimoll db
DOIRQSBPFJWKBE-UHFFFAOYSA-N
1,2-dibutyl benzene-1,2-dicarboxylate
3026
278.35
84-74-2
1.045
171°C (340°F)
C16H22O4
MFCD00009441
UN3082
14,3035
Soluble in water. (0.4g/L)
CCCCOC(=O)C1=CC=CC=C1C(=O)OCCCC
278.35
CHEBI:34687
99%

Chemical Identifiers

84-74-2
278.35
DOIRQSBPFJWKBE-UHFFFAOYSA-N
3026
1,2-dibutyl benzene-1,2-dicarboxylate
C16H22O4
MFCD00009441
dibutyl phthalate, di-n-butyl phthalate, n-butyl phthalate, butyl phthalate, celluflex dpb, elaol, genoplast b, palatinol c, polycizer dbp, unimoll db
CHEBI:34687
CCCCOC(=O)C1=CC=CC=C1C(=O)OCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P260-P271-P281-P304+P340-P308+P313-P501c

H336-H360Df-H373

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 201-557-4

RTECSNumber : TI0875000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only