missing translation for 'onlineSavingsMsg'
Learn More

Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] Chloride 97.0+%, TCI America™

Supplier:  TCI America D25425G

 View more versions of this product

Catalog No. D25425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Di-mu-hydroxo-bis[(N,N,N′,N′-tetramethylethylenediamine)copper(II)] Chloride
Black-Gray
MFCD01321163
cu-tmeda catalyst, di-mu-hydroxo-bis n,n,n'.n'-tetramethylethylenediamine copper ii chloride, cu-tmedacatalyst, bis chlorocopperol ; bis temed, di-hydroxo-bis n,n,n',n'-tetramethylethylenediamine copper ii chloride, chlorohydroxo n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine copper ii, copper ii chloride hydroxide n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine complex dimer, di-, i-hydroxo-bis n,n,n inverted exclamation marka,n inverted exclamation marka-tetramethylethylenediamine copper ii chloride, i-hydroxo-bis n,n,n inverted exclamation marka.n inverted exclamation marka-tetramethylethylenediamine copper ii chloride
O.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C
466.44
464.42
Crystalline Powder
30698-64-7
C12H36Cl2Cu2N4O2
5 g
VVXKYYDFGPZSOZ-UHFFFAOYSA-L
bis([2-(dimethylamino)ethyl]dimethylamine) bis(chlorocopper) dihydrate
11190467
≥97.0% (T)

Chemical Identifiers

30698-64-7
466.44
VVXKYYDFGPZSOZ-UHFFFAOYSA-L
11190467
O.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C
C12H36Cl2Cu2N4O2
MFCD01321163
cu-tmeda catalyst, di-mu-hydroxo-bis n,n,n'.n'-tetramethylethylenediamine copper ii chloride, cu-tmedacatalyst, bis chlorocopperol ; bis temed, di-hydroxo-bis n,n,n',n'-tetramethylethylenediamine copper ii chloride, chlorohydroxo n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine copper ii, copper ii chloride hydroxide n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine complex dimer, di-, i-hydroxo-bis n,n,n inverted exclamation marka,n inverted exclamation marka-tetramethylethylenediamine copper ii chloride, i-hydroxo-bis n,n,n inverted exclamation marka.n inverted exclamation marka-tetramethylethylenediamine copper ii chloride
bis([2-(dimethylamino)ethyl]dimethylamine) bis(chlorocopper) dihydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions