missing translation for 'onlineSavingsMsg'
Learn More

Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] Chloride 97.0+%, TCI America™

Catalog No. p-7127358
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25425G 5 g
D254225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25425G Supplier TCI America Supplier No. D25425G
Only null left

Chemical Identifiers

CAS 30698-64-7
Molecular Formula C12H36Cl2Cu2N4O2
Molecular Weight (g/mol) 466.44
MDL Number MFCD01321163
InChI Key VVXKYYDFGPZSOZ-UHFFFAOYSA-L
Synonym cu-tmeda catalyst, di-mu-hydroxo-bis n,n,n'.n'-tetramethylethylenediamine copper ii chloride, cu-tmedacatalyst, bis chlorocopperol ; bis temed, di-hydroxo-bis n,n,n',n'-tetramethylethylenediamine copper ii chloride, chlorohydroxo n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine copper ii, copper ii chloride hydroxide n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine complex dimer, di-, i-hydroxo-bis n,n,n inverted exclamation marka,n inverted exclamation marka-tetramethylethylenediamine copper ii chloride, i-hydroxo-bis n,n,n inverted exclamation marka.n inverted exclamation marka-tetramethylethylenediamine copper ii chloride
PubChem CID 11190467
IUPAC Name bis([2-(dimethylamino)ethyl]dimethylamine) bis(chlorocopper) dihydrate
SMILES O.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C

Specifications

Color Black-Gray
Quantity 5 g
Formula Weight 464.42
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Di-mu-hydroxo-bis[(N,N,N′,N′-tetramethylethylenediamine)copper(II)] Chloride