missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] Chloride 97.0+%, TCI America™
Supplier: TCI America D254225G
Specifications
| Di-mu-hydroxo-bis[(N,N,N′,N′-tetramethylethylenediamine)copper(II)] Chloride | |
| Black-Gray | |
| MFCD01321163 | |
| cu-tmeda catalyst, di-mu-hydroxo-bis n,n,n'.n'-tetramethylethylenediamine copper ii chloride, cu-tmedacatalyst, bis chlorocopperol ; bis temed, di-hydroxo-bis n,n,n',n'-tetramethylethylenediamine copper ii chloride, chlorohydroxo n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine copper ii, copper ii chloride hydroxide n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine complex dimer, di-, i-hydroxo-bis n,n,n inverted exclamation marka,n inverted exclamation marka-tetramethylethylenediamine copper ii chloride, i-hydroxo-bis n,n,n inverted exclamation marka.n inverted exclamation marka-tetramethylethylenediamine copper ii chloride | |
| O.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C | |
| 466.44 | |
| 464.42 | |
| Crystalline Powder |
| 30698-64-7 | |
| C12H36Cl2Cu2N4O2 | |
| 25 g | |
| VVXKYYDFGPZSOZ-UHFFFAOYSA-L | |
| bis([2-(dimethylamino)ethyl]dimethylamine) bis(chlorocopper) dihydrate | |
| 11190467 | |
| ≥97.0% (T) |
Chemical Identifiers
| 30698-64-7 | |
| 466.44 | |
| VVXKYYDFGPZSOZ-UHFFFAOYSA-L | |
| 11190467 | |
| O.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C |
| C12H36Cl2Cu2N4O2 | |
| MFCD01321163 | |
| cu-tmeda catalyst, di-mu-hydroxo-bis n,n,n'.n'-tetramethylethylenediamine copper ii chloride, cu-tmedacatalyst, bis chlorocopperol ; bis temed, di-hydroxo-bis n,n,n',n'-tetramethylethylenediamine copper ii chloride, chlorohydroxo n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine copper ii, copper ii chloride hydroxide n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine complex dimer, di-, i-hydroxo-bis n,n,n inverted exclamation marka,n inverted exclamation marka-tetramethylethylenediamine copper ii chloride, i-hydroxo-bis n,n,n inverted exclamation marka.n inverted exclamation marka-tetramethylethylenediamine copper ii chloride | |
| bis([2-(dimethylamino)ethyl]dimethylamine) bis(chlorocopper) dihydrate |
Safety and Handling
TSCA : No