missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-mu-chlorobis[5-chloro-2-[(4-chlorophenyl)(hydroxyimino)methyl]phenyl]palladium(II) Dimer 95.0+%, TCI America™
$180.81 - $577.86
Chemical Identifiers
| CAS | 287410-78-0 |
|---|---|
| Molecular Formula | C26H18Cl6N2O2Pd2 |
| Molecular Weight (g/mol) | 815.982 |
| MDL Number | MFCD09038424 |
| InChI Key | QPUPWYGEQHWYOF-INIQRQEMSA-N |
| Synonym | Najera Catalyst I |
| PubChem CID | 132774960 |
| IUPAC Name | (NE)-N-[(4-chlorobenzene-6-id-1-yl)-(4-chlorophenyl)methylidene]hydroxylamine;dichloroniopalladium;palladium |
| SMILES | C1=CC(=CC=C1C(=NO)C2=CC=C(C=[C-]2)Cl)Cl.C1=CC(=CC=C1C(=NO)C2=CC=C(C=[C-]2)Cl)Cl.[ClH+][Pd][ClH+].[Pd] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D3806250MG
|
TCI America
D3806250MG |
250 mg |
Each for $180.81
|
|
|||||
|
D38061G
|
TCI America
D38061G |
1 g |
Each for $577.86
|
|
|||||
Chemical Identifiers
| 287410-78-0 | |
| 815.982 | |
| QPUPWYGEQHWYOF-INIQRQEMSA-N | |
| 132774960 | |
| C1=CC(=CC=C1C(=NO)C2=CC=C(C=[C-]2)Cl)Cl.C1=CC(=CC=C1C(=NO)C2=CC=C(C=[C-]2)Cl)Cl.[ClH+][Pd][ClH+].[Pd] |
| C26H18Cl6N2O2Pd2 | |
| MFCD09038424 | |
| Najera Catalyst I | |
| (NE)-N-[(4-chlorobenzene-6-id-1-yl)-(4-chlorophenyl)methylidene]hydroxylamine;dichloroniopalladium;palladium |
Specifications
| 287410-78-0 | |
| C26H18Cl6N2O2Pd2 | |
| 250 mg | |
| QPUPWYGEQHWYOF-INIQRQEMSA-N | |
| (NE)-N-[(4-chlorobenzene-6-id-1-yl)-(4-chlorophenyl)methylidene]hydroxylamine;dichloroniopalladium;palladium | |
| 132774960 | |
| ≥95.0% (T) | |
| Di-mu-chlorobis[5-chloro-2-[(4-chlorophenyl)(hydroxyimino)methyl]phenyl]palladium(II) Dimer |
| Yellow | |
| MFCD09038424 | |
| Najera Catalyst I | |
| C1=CC(=CC=C1C(=NO)C2=CC=C(C=[C-]2)Cl)Cl.C1=CC(=CC=C1C(=NO)C2=CC=C(C=[C-]2)Cl)Cl.[ClH+][Pd][ClH+].[Pd] | |
| 815.982 | |
| 813.97 | |
| Crystalline Powder |
Safety and Handling
TSCA : No