missing translation for 'onlineSavingsMsg'
Learn More

Di-^m-chlorodichlorobis(ethylene)diplatinum(II), 97%

Catalog No. AA39285MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39285MD 250 mg
AA3928503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA39285MD Supplier Thermo Scientific Chemicals Supplier No. 039285MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

CAS: 12073-36-8 | C4H8Cl4Pt2 | 588.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12073-36-8
Molecular Formula C4H8Cl4Pt2
Molecular Weight (g/mol) 588.08
MDL Number MFCD00058713
InChI Key XBLBDYCZFZESPN-UHFFFAOYSA-J
Synonym di-mu-chloro-dichlorobis ethylene diplatinum ii, zeise's dimer, ptcl2 c2h4 2, di-chloro-dichlorobis ethylene diplatinum ii, di-micro-chlorodichlorobis ethylene diplatinum ii
PubChem CID 11124780
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Pt++].[Pt++].C=C.C=C

Specifications

Quantity 250 mg
IUPAC Name dichloroplatinum;ethene
Formula Weight 588.1
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Di-mu-chlorodichlorobis(ethylene)diplatinum(II)
Hazard Category H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
EINECSNumber 235-135-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only