CAS RN 12073-36-8
IUPAC Name:
diplatinum(2+) bis(ethene) tetrachloride
Synonyms:
diplatinum(2+) bis(ethylene) tetrachloride
Molecular Weight (g/mol):
588.08
Molecular Formula:
C4H8Cl4Pt2
InChi Key:
XBLBDYCZFZESPN-UHFFFAOYSA-J
SMILES:
[Cl-].[Cl-].[Cl-].[Cl-].[Pt++].[Pt++].C=C.C=C
1
–
1
of
1
results
1
Di-^m-chlorodichlorobis(ethylene)diplatinum(II), 97%
CAS: 12073-36-8 Molecular Formula: C4H8Cl4Pt2 Molecular Weight (g/mol): 588.08 MDL Number: MFCD00058713 InChI Key: XBLBDYCZFZESPN-UHFFFAOYSA-J Synonym: di-mu-chloro-dichlorobis ethylene diplatinum ii,zeise's dimer,ptcl2 c2h4 2,di-chloro-dichlorobis ethylene diplatinum ii,di-micro-chlorodichlorobis ethylene diplatinum ii PubChem CID: 11124780 SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Pt++].[Pt++].C=C.C=C