missing translation for 'onlineSavingsMsg'
Learn More

Di-2-thienyl ketone, 98%

Catalog No. AAA1206206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1206206 5 g
AAA1206203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1206206 Supplier Thermo Scientific Chemicals Supplier No. A1206206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 704-38-1 | C9H6OS2 | 194.266 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 704-38-1
Molecular Formula C9H6OS2
Molecular Weight (g/mol) 194.266
MDL Number MFCD00052096
InChI Key GUTQMBQKTSGBPQ-UHFFFAOYSA-N
Synonym bis 2-thienyl ketone, 2-thienyl ketone, di thiophen-2-yl methanone, methanone, di-2-thienyl, di-2-thienylmethanone, di-2-thienyl ketone, di-thiophen-2-yl-methanone, 2-2-thenoyl thiophene, bis thiophen-2-yl methanone
PubChem CID 69713
IUPAC Name dithiophen-2-ylmethanone
SMILES C1=CSC(=C1)C(=O)C2=CC=CS2

Specifications

Melting Point 87°C to 90°C
Quantity 5 g
Beilstein 129168
Formula Weight 194.28
Percent Purity 98%
Chemical Name or Material Di-2-thienyl ketone
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No

RUO – Research Use Only