CAS RN 704-38-1
IUPAC Name:
bis(thiophen-2-yl)methanone
Synonyms:
di-thiophen-2-yl-methanone
Molecular Weight (g/mol):
194.27
Molecular Formula:
C9H6OS2
InChi Key:
GUTQMBQKTSGBPQ-UHFFFAOYSA-N
SMILES:
O=C(C1=CC=CS1)C1=CC=CS1
1
–
1
of
1
results
1
Promotions Available
Di-2-thienyl ketone, 98%
Di-2-thienyl ketone, 98%
CAS: 704-38-1 Molecular Formula: C9H6OS2 Molecular Weight (g/mol): 194.266 MDL Number: MFCD00052096 InChI Key: GUTQMBQKTSGBPQ-UHFFFAOYSA-N Synonym: bis 2-thienyl ketone,2-thienyl ketone,di thiophen-2-yl methanone,methanone, di-2-thienyl,di-2-thienylmethanone,di-2-thienyl ketone,di-thiophen-2-yl-methanone,2-2-thenoyl thiophene,bis thiophen-2-yl methanone PubChem CID: 69713 IUPAC Name: dithiophen-2-ylmethanone SMILES: C1=CSC(=C1)C(=O)C2=CC=CS2