missing translation for 'onlineSavingsMsg'
Learn More

Di-2-pyridyl thionocarbonate, 98%

Catalog No. p-7026453
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2245103 1 g
AAB2245106 5 g
AAB2245114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2245103 Supplier Thermo Scientific Chemicals Supplier No. B2245103
Only null left

CAS: 96989-50-3 | C11H8N2O2S | 232.257 g/mol

Di-2-pyridyl thionocarbonate is used in the synthesis of 10-des(carbamoyloxy)-10-isothiocyanatoporfiromycin. It is also used as a substitute of thiophosgene, in the preparation of isothiocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-2-pyridyl thionocarbonate is used in the synthesis of 10-des(carbamoyloxy)-10-isothiocyanatoporfiromycin. It is also used as a substitute of thiophosgene, in the preparation of isothiocyanate.

Solubility
Soluble in water 9280 g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 96989-50-3
Molecular Formula C11H8N2O2S
Molecular Weight (g/mol) 232.257
MDL Number MFCD00074870
InChI Key IKYOVSVBLHGFMA-UHFFFAOYSA-N
Synonym di-2-pyridyl thionocarbonate, o,o-di pyridin-2-yl carbonothioate, o,o'-di-2-pyridyl thiocarbonate, o,o-dipyridin-2-yl carbonothioate, di 2-pyridyl thionocarbonate, carbonothioic acid, o,o-di-2-pyridinyl ester, di 2-pyridyloxy methane-1-thione, o,o-dipyridin-2-yl thiocarbonate, bis pyridin-2-yloxy methanethione, di-2-pyridyl thiocarbomate
PubChem CID 719784
IUPAC Name dipyridin-2-yloxymethanethione
SMILES C1=CC=NC(=C1)OC(=S)OC2=CC=CC=N2

Specifications

Melting Point 96°C to 99°C
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water 9280g/L).
Formula Weight 232.26
Percent Purity 98%
Chemical Name or Material Di-2-pyridyl thionocarbonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only