missing translation for 'onlineSavingsMsg'
Learn More

Di(2-ethylhexyl)amine 98.0+%, TCI America™

Catalog No. D051025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D051025ML 25 mL
D0510500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D051025ML Supplier TCI America Supplier No. D051025ML

Chemical Identifiers

CAS 106-20-7
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.463
MDL Number MFCD00009489
InChI Key SAIKULLUBZKPDA-UHFFFAOYSA-N
Synonym bis 2-ethylhexyl amine, di 2-ethylhexyl amine, diisooctylamine, 1-hexanamine, 2-ethyl-n-2-ethylhexyl, bis-2-ethylhexylamine, 2,2'-diethyldihexylamine, 2,2'-diethylhexylamine, di-2-ethylhexyl amine, dihexylamine, 2,2'-diethyl, ccris 4619
PubChem CID 7791
IUPAC Name 2-ethyl-N-(2-ethylhexyl)hexan-1-amine
SMILES CCCCC(CC)CNCC(CC)CCCC

Specifications

Color Yellow
Boiling Point 123°C
Quantity 25 mL
UN Number 2922
Formula Weight 241.46
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Di(2-ethylhexyl)amine
EINECSNumber (2)-0176&(2)-0133
RTECSNumber IH6825000
TSCA Yes