accessibility menu, dialog, popup

UserName

CAS RN 106-20-7

H3CCH3HNCH3CH3

CAS RN 106-20-7

IUPAC Name: bis(2-ethylhexyl)amine
Synonyms: di-(2-ethylhexyl)amine
Molecular Weight (g/mol): 241.46
Molecular Formula: C16H35N
InChi Key: SAIKULLUBZKPDA-UHFFFAOYNA-N
SMILES: CCCCC(CC)CNCC(CC)CCCC
Boiling Point 
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)