missing translation for 'onlineSavingsMsg'
Learn More

Di-1-naphthylmethanol 98.0+%, TCI America™

Catalog No. D37505G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D37505G Supplier TCI America Supplier No. D37505G
Only null left

Chemical Identifiers

CAS 62784-66-1
Molecular Formula C21H16O
Molecular Weight (g/mol) 284.36
MDL Number MFCD11096937
InChI Key NHIXQVYVFRYTOB-UHFFFAOYSA-N
Synonym Bis(alpha-naphthyl)methanol
PubChem CID 593473
IUPAC Name bis(naphthalen-1-yl)methanol
SMILES OC(C1=CC=CC2=CC=CC=C12)C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 148°C
Quantity 5 g
Formula Weight 284.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Di-1-naphthylmethanol