accessibility menu, dialog, popup

UserName

OH

CAS RN 62784-66-1

IUPAC Name: bis(naphthalen-1-yl)methanol
Synonyms: bis(naphthalen-1-yl)methanol
Molecular Weight (g/mol): 284.36
Molecular Formula: C21H16O
InChi Key: NHIXQVYVFRYTOB-UHFFFAOYSA-N
SMILES: OC(C1=CC=CC2=CC=CC=C12)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results