missing translation for 'onlineSavingsMsg'
Learn More

DHX38 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501558634
Encompass
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501558634 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501558634 Supplier Bethyl Laboratories Inc Supplier No. A300859AT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict

Probable ATP-binding RNA helicase involved in pre-mRNA splicing.

Specifications

Antigen DHX38
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene DHX38
Gene Accession No. Q92620
Gene Alias ATP-dependent RNA helicase DHX38, DDX38, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 38, DEAH (Asp-Glu-Ala-His) box polypeptide 38, DEAH box protein 38, hPrp16, KIAA0224, pre-mRNA splicing factor ATP-dependent RNA helicase PRP16, PRP16, PRP16 homolog of S.cerevisiae, PRPF16
Gene Symbols Dhx38
Host Species Rabbit
Immunogen Between 1200 and C-term
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64340, 9785
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less