missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DHFR Recombinant Rabbit Monoclonal Antibody (23GB3575)
GREENER_CHOICE

Catalog No. MA553398
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553398 100 μL
MA553399 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553398 Supplier Invitrogen™ Supplier No. MA553398
Only null left

Rabbit Recombinant Monoclonal Antibody

DHFR Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Dihydrofolate reductase converts dihydrofolate into tetrahydrofolate, and is required for the synthesis of purines, thymidylic acid and certain amino acids. While the functional dihydrofolate reductase gene has been mapped to chromosome 5, multiple intronless processed pseudogenes or dihydrofolate reductase like genes have been identified on separate chromosomes. Dihydrofolate reductase deficiency has been linked to megaloblastic anemia.

Specifications

Antigen DHFR
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3575
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DHFR
Gene Accession No. P00374, P00375, Q920D2
Gene Alias 8430436I03Rik; AA607882; AI662710; AW555094; Dhfr; Dhfr1; DHFRP1; Dihydrofolate reductase; Dihydrofolate reductase 1 (active); DYR
Gene Symbols DHFR
Host Species Rabbit
Immunogen A synthetic peptide of human DHFR (134-187AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13361, 1719, 24312
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less