missing translation for 'onlineSavingsMsg'
Learn More

Medisca DHEA (Prasterone) (Micronized)

Catalog No. 14186216
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186216 25 g
14186103 1 kg
14185455 5 g
14185505 100 g
14186203 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14186216 Supplier Medisca Supplier No. 073304

DHEA (Micronized) (Prasterone) is a white to almost white crystalline powder. Particle size distribution: 90% ≤ 50 μm.

DHEA (Micronized) (Prasterone) is a white to almost white crystalline powder. Particle size distribution: 90% ≤ 50 μm.

Alternative name(s): Dehydroepiandrosterone, dehydroisoandrosterone

Specifications

Chemical Name or Material DHEA (Prasterone) (Micronized)
Packaging Jar 300ml, White, HDPE, Round, 53-400, TRIVENI-DB, CT
CAS 53-43-0
Type Drug Standard
Molecular Weight (g/mol) 288.4282
Quantity 25 g
Percent Purity 97.5 to 102 %
Grade Ungraded