missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DH5α Competent Cells; High Efficiency

Catalog No. FEREC0112
Change view
Click to view available options
Quantity:
20 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREC0112 20 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREC0112 Supplier Thermo Scientific™ Supplier No. EC0112

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High efficiency, chemically competent E. coli cells ideal for construction of gene banks or generation of cDNA libraries using plasmid-derived vectors

  • The φ80dlacZΔM15 marker provides α-complementation of the β-galactosidase gene from pUC or similar vectors to allow blue/white colony screening on bacterial agar plates containing Bluo-Gal or X-Gal
  • RecA1 and endA1 mutations in DH5α cells improve insert stability and quality of the extracted plasmid DNA
  • High transformation efficiency: >1x109 cfu/μg pUC19 DNA
  • Suitable for routine and high-throughput cloning applications
  • Genetic markers that allow for blue/white colony screening
  • Convenient two reactions per vial packaging
  • S.O.C. Outgrowth medium included
  • DH5α Competent Cells are suitable for variety of applications requiring high transformation efficiency:
    • Efficient DNA cloning derived from PCR, cDNA-generation reactions
    • recA1 marker facilitates working with difficult to transform DNA
    • Ideal for generation of cDNA libraries using plasmid-derived vectors
    • Site-directed mutagenesis
  • Genotype: F– φ80lacZΔ M15 Δ (lacZYA-argF) U169 recA1 endA1 hsdR17 (rK– mK+) phoA supE44 λ- thi–1 gyrA96 relA1

Specifications

Description Chemically Competent
Contains F' Episome Lacks F' Episome
Content And Storage Dry Ice (-80°C)
Bacterial or Yeast Strain DH5α
Species E. coli
Format Tube
For Use With (Application) Cloning
Quantity 20 Reactions