missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DH10B Competent Cells; High Efficiency

Catalog No. FEREC0113
Change view
Click to view available options
Quantity:
20 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREC0113 20 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREC0113 Supplier Thermo Scientific™ Supplier No. EC0113

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High efficiency, chemically competent E. coli cells. The DH10B strain is suitable for cloning DNA containing methylcytosine, 5-hydroxymethylcytosine, and methyladenine, allowing both prokaryotic and eukaryotic genomic DNA to be cloned efficiently.

  • Suitable for cloning DNA containing methylcytosine, 5-hydroxymethylcytosine, and methyladenine, allowing both prokaryotic and eukaryotic genomic DNA to be cloned efficiently
  • These cells are ideal for the construction of gene banks or for the generation of cDNA libraries using plasmid-derived vectors
  • The φ80dlacZΔM15 marker provides α-complementation of the β-galactosidase gene from pUC or similar vectors to allow blue/white colony screening on bacterial agar plates containing Bluo-Gal or X-Gal
  • High transformation efficiency: >1x109 cfu/μg pUC19 DNA
  • Suitable for routine and high-throughput cloning applications
  • Allows for blue/white colony screening
  • Convenient two reactions per vial packaging
  • S.O.C. Outgrowth medium included
  • Suitable for variety of applications requiring high transformation efficiency:
    • Efficient DNA cloning both of prokaryotic and eukaryotic origin
    • Cloning of large plasmids and BACs
    • Ideal for generation of cDNA libraries using plasmid-derived vectors
    • Site directed mutagenesis
  • Genotype: F– mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 endA1 araD139 Δ (ara-leu)7697 galU galK λ– rpsL(StrR) nupG

Note: Optimized for use with the CloneJET PCR Cloning Kit (Cat. Nos. K1231, K1232) and Phusion Site-Directed Mutagenesis Kit (Cat. No. F541), and available as combo versions (Cat. Nos. K123120, K123240, F542)

Specifications

Description Chemically Competent
Contains F' Episome Lacks F' Episome
Content And Storage Dry Ice (-80°C)
Bacterial or Yeast Strain DH10B
Species E. coli
Format Tube
For Use With (Application) Cloning
Quantity 20 Reactions