missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ dGTP BigDye™ Terminator v3.0 Ready Reaction Cycle Sequencing Kit

Catalog No. 4390229
Encompass
Change view
Click to view available options
Quantity:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4390229 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4390229 Supplier Applied Biosystems™ Supplier No. 4390229

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Contains the AmpliTaq™ DNA Polymerase, FS, BigDye terminators v3.0, pGEM™ - 3Zf(+) Control Template, -21 M13 Control Primer (forward), and Premixed Ready Reaction.

The dGTP BigDye™ terminator cycle sequencing kit's fluorescent dideoxy terminator chemistry is optimized for sequencing through GT- and G-rich templates.

  • Ensure reliable results with difficult regions and finishing applications
  • Sequence templates with challenging sequence contexts
  • Sequence problematic templates that standard chemistries cannot
  • The dGTP BigDye™ Terminator Kit replaces dITP with dGTP and it enables you to extend through those difficult-to-sequence regions with confidence, avoiding early signal loss in these samples
  • Optimized for GT-Rich templates: Obtain higher success rate even with your difficult GT-rich templates
  • By using the same chemistries as the standard BigDye™ Terminator Kits (only replacing dITP with dGTP) the dGTP BigDye™ Terminator Kit provides proven performance
  • Optimized for GT-rich and other difficult-to-sequence templates

Order Info

100 rxns

Specifications

Content And Storage The kit contains the AmpliTaq™ DNA Polymerase, FS, BigDye
terminators v3.0, pGEM™ - 3Zf(+) Control Template,
-21 M13 Control Primer (forward), and Premixed Ready Reaction.
Store kit at -15 to -25°C.
Format 12-well Strips, 8-well Strip, 96- or 384-well Plate
For Use With (Application) GT-Rich or Difficult Template Sequencing
No. of Reactions 100 reactions
Product Line BigDye Terminator, BigDye
Product Type Terminator V3.0 Ready Reaction Cycle Sequencing Kit
Quantity 100 reactions
Shipping Condition Dry Ice
Technique Fluorescent Dye Terminator Sequencing
Template Compatibility BAC DNA, Genomic DNA (Bacterial), Plasmid DNA (≤15Kb), PCR Amplicons, Single Stranded DNA
What information should I include when submitting data to Genetic Analysis Technical Support?

When contacting Genetic Analysis Support (FAS, FSE, Technical Support), please have (if possible):

Instrument model and serial numbers
Version of the software are you using
Application and/or kit information
Kit lot number
Number of runs on the capillary or array
Polymer type and lot number
Buffer lot number
Hi-Di Formamide lot number or loading solution information
Sample data that can be sent to the support person
Details on:
  -Reaction setup (i.e., how much DNA was used, primer, etc.)
  -How the reactions were cleaned up (alcohol precipitation, columns, etc)
  -What template DNA was used (i.e. plasmid, PCR product)
  -How the template DNA was prepared?

All of this information will help the support person quickly and accurately assess the problem and provide you with recommendations for resolution.

I have a long homopolymer stretch in my sample that I cannot get through. Do you have any tips for getting through it?

In order to get through homopolymeric regions, you can either use anchored sequencing primers or try using the dRhodamine Terminator Cycle Sequencing Kit since it uses ddTTP instead of ddUTP and has been shown to be less prone to producing stutters, specifically with poly-T regions.

For more information on sequencing through homopolymer stretches, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

I am trying to sequence GC-rich DNA without success. Do you have any tips for getting through this?

When trying to sequence through GC-rich regions, the following tips may improve your success:

• Set the reaction up at 1X or 0.5X strength. Heavily diluting the BigDye Terminator Ready Reaction mix can reduce the success of the sequecing reaction through long GC stretches.

• Perform a hot start at 98–99°C for 5 minutes.

• Use 5% (w/v) DMSO or freshly made betaine in the reaction.

• Use the dGTP BigDye Terminator Cycle Sequencing Kit. G peaks will appear compressed due to the presence of ddGTP, but sequencing through long GC stretches using the dGTP kit is typically more successful than with the standard BigDye Terminator Cycle Sequencing Kits (containing ddITP). For more information on sequencing GC-rich DNA, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

My DNA sequencing data from the capillary electrophoresis system does not look good. Why?

Capillary electrophoresis systems are complex systems, and troubleshooting data quality can be complicated. The biggest contributors to data quality can be divided into these areas: software, chemistry, and instrument performance.

Troubleshooting software
Ensure that the run module you are using and the dye set match the chemistry and instrument setup. For example, if you are running a multicapillary instrument with a 50 cm array on it, the run module should have a “50” in the name. On a multicapillary system, BigDye Terminator v3.1 should run under Dye Set Z, BigDye Terminator v1.1 chemistry should run in Dye Set E (on the ABI PRISM 310 Genetic Analyzer, both chemistries run under Filter Set E, but require different matrices for analysis). The mobility file (DyeSet/Primer) should have the correct instrument, polymer, and chemistry in the name (e.g. KB_3130_POP7_BDTv.3.mob).

Troubleshooting chemistry and instrument performance
•Sequencing Standards: validate instrument
If Sequencing Standards fail, it suggests a possible electrophoresis problem due to array, polymer, buffer, water, septa, plastics, or maybe a hardware failure such as autosampler, laser, camera, etc.

•pREF-BDT Control DNA: validate cycle sequencing and its clean-up
If the pREF-BDT control reactions fail and the Sequencing Standards pass, look into potential problems with the sequencing kit, thermal cycler, and cleanup procedure.
The pREF-BDT Control DNA and M13 (–21) primers needed to run the reactions are included with each BigDye Terminator Cycle Sequencing Kit

•Custom Internal Controls: validate PCR and its clean-up
If the Sequencing Standards and the pGEM controls passed, then look into potential problems with template quantity and quality, primers, PCR reaction and purification.

For more information on troubleshooting your data, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: 3rd Edition, which can be downloaded from this link (https://www.thermofisher.com/us/en/home/global/forms/sanger-sequencing-guide-download-form.html).

What are the concentrations of the pREF-BDT and M13 primer controls that come with the BigDye Terminator kits?

The concentration of the pREF-BDT control is 200 ng/µL. The concentration of the M13 primer is 0.8 pmol/µL.

What instruments can the dGTP BigDye Terminator Cycle Sequencing Kit be run on?

The dGTP BigDye Terminator Cycle Sequencing Kit can be run on all of our capillary electrophoresis platforms:

- Applied Biosystems 3500/3500xL Genetic Analyzers
- Applied Biosystems 3730/3730xl DNA Analyzers
- Applied Biosystems 310 Genetic Analyzer
- Applied Biosystems 3130/3130xl DNA Analyzers

However, due to the nature of the chemistry, compressions in the data are expected.

When should I use the dGTP BigDye Terminator Cycle Sequencing Kit?

The dGTP BigDye Terminator Cycle Sequencing Kit should be used for difficult templates, such as templates that are GT-rich, G-rich templates or are difficult to sequence with other kits. They should be used only when you cannot obtain good data using standard BigDye Terminator kits.

What is the dGTP BigDye Terminator Cycle Sequencing Kit?

The dGTP BigDye Terminator Cycle Sequencing Kits are kits that can be used for sequencing difficult templates where early signal loss may occur. The kit is not recommended for routine sequencing as the kit uses dGTP instead of dITP and compressions can occur with the data.

Can I purchase the pREF-BDT control from the BigDye Terminator kit separately?

We do not sell the pREF-BDT control separately from the DNA sequencing kit.

What is the control in the BigDye Terminator kit?

The control is pREF-BDT control plasmid DNA which serves as a double-stranded control


For Research Use Only. Not for use in diagnostic procedures.