missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DGKZ Monoclonal Antibody (K49027_8F3), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA555685

 View more versions of this product

Catalog No. MA555685


Only null left

Description

Description

Sequence of this protein is as follows: STGSKDVVRY LLDHAPPEIL DAVEENGETC LHQAAALGQR TICHYIVEAG ASLMKTDQQG DTPRQRAEKA QDTELAAYLE NRQHYQMIQR

The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It may attenuate protein kinase C activity by regulating diacylglycerol levels in intracellular signaling cascade and signal transduction. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified.
Specifications

Chemical Identifiers

DGKZ
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
104 kDa diacylglycerol kinase; DAG kinase DAGK6; DAG kinase zeta; DAGK5; DAGK6; DGK-IV; DGKZ; DGK-zeta; Diacylglycerol kinase zeta; diacylglycerol kinase, zeta 104kDa; Diglyceride kinase; diglyceride kinase zeta; E130307B02Rik; ENSMUSG00000075035; F730209L11Rik; hDGKzeta; mDGK[z]
Recombinant human DGKZ (immunogen range is 832-921).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
DGKZ
K49027_8F3
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Mouse
Q13574, Q80UP3
104418, 8525
IgG1
Protein A
Antibody

Specifications

DGKZ
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Q13574, Q80UP3
DGKZ
Recombinant human DGKZ (immunogen range is 832-921).
50 μL
Primary
Human, Mouse
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
K49027_8F3
Unconjugated
DGKZ
104 kDa diacylglycerol kinase; DAG kinase DAGK6; DAG kinase zeta; DAGK5; DAGK6; DGK-IV; DGKZ; DGK-zeta; Diacylglycerol kinase zeta; diacylglycerol kinase, zeta 104kDa; Diglyceride kinase; diglyceride kinase zeta; E130307B02Rik; ENSMUSG00000075035; F730209L11Rik; hDGKzeta; mDGK[z]
Mouse
Protein A
RUO
104418, 8525
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions