missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DGKZ Monoclonal Antibody (K49027_10E9), Invitrogen™

Catalog No. MA556067
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556067 200 μL
MA555687 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556067 Supplier Invitrogen™ Supplier No. MA556067

Mouse Monoclonal Antibody

Sequence of this protein is as follows: STGSKDVVRY LLDHAPPEIL DAVEENGETC LHQAAALGQR TICHYIVEAG ASLMKTDQQG DTPRQRAEKA QDTELAAYLE NRQHYQMIQR

The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It may attenuate protein kinase C activity by regulating diacylglycerol levels in intracellular signaling cascade and signal transduction. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified.

Specifications

Antigen DGKZ
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K49027_10E9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene DGKZ
Gene Accession No. Q13574, Q80UP3
Gene Alias 104 kDa diacylglycerol kinase; DAG kinase DAGK6; DAG kinase zeta; DAGK5; DAGK6; DGK-IV; DGKZ; DGK-zeta; Diacylglycerol kinase zeta; diacylglycerol kinase, zeta 104kDa; Diglyceride kinase; diglyceride kinase zeta; E130307B02Rik; ENSMUSG00000075035; F730209L11Rik; hDGKzeta; mDGK[z]
Gene Symbols DGKZ
Host Species Mouse
Immunogen Recombinant human DGKZ (immunogen range is 832-921).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104418, 8525
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less