missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DGKH Recombinant Mouse Monoclonal Antibody (K94091_8F5)
GREENER_CHOICE

Catalog No. MA568022
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA568022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA568022 Supplier Invitrogen™ Supplier No. MA568022
Only null left

Mouse Recombinant Monoclonal Antibody

DGKH Recombinant Monoclonal Antibody for Western Blot

This gene encodes a member of the diacylglycerol kinase (DGK) enzyme family. Members of this family are involved in regulating intracellular concentrations of diacylglycerol and phosphatidic acid. Variation in this gene has been associated with bipolar disorder. Alternatively spliced transcript variants have been identified.

Specifications

Antigen DGKH
Applications Western Blot
Classification Recombinant Monoclonal
Clone K94091_8F5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene Dgkh
Gene Accession No. Q86XP1
Gene Alias 5930402B05Rik; D130015C16; DAG kinase eta; DGK; DGKeta; DGK-eta; DGKH; Diacylglycerol kinase eta; diacylglycerol kinase, eta; diglyceride kinase eta; DKFZp761I1510; EC 2.7.1.107; RGD1561955; si:ch211-93a19.1
Gene Symbols Dgkh
Host Species Mouse
Immunogen Recombinant human DGKH
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160851
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less