Learn More
Applied Biosystems™ dGdmf, 1 μmol ABI 3900 HT Columns
Catalog No. 4316681
Encompass
Change view
Click to view available options
Quantity:
200 Columns
Catalog No. Quantity
4316681 200 Columns
1 options
Catalog No. 4316681 Supplier Applied Biosystems™ Supplier No. 4316681
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The 1μmol dG(dmf) ABI 3900 HT columns are fully assembled, low-cost columns that produce high-quality oligonucleotides. These supports are filled with polystryene, a material which has several advantages over the more conventional, controlled pore glass (CPG) filled columns.
Designed for PerformanceThe greater hydrophobicity of polystyrene results in a higher yield of full-length product. Side reactions, such as extraneous chain growth, are minimized by the inert nature of this support surface.
  • Proprietary design and manufacturing methods ensure consistency and high quality to give you optimal synthesis quality.
  • Hydrophobic, polystyrene-filled columns sustain an anhydrous environment to promote efficient coupling while minimizing nonspecific binding sites, yielding an efficient synthesis.
  • Polystyrene support allows synthesis of oligonucleotides of a variety of lengths, giving you flexibility in high-throughput oligonucleotide synthesis.
Specifications
Packaging Type Shipped at Room Temperature
Quantity 200 Columns
For Research Use Only. Not for use in diagnostic procedures.