missing translation for 'onlineSavingsMsg'
Learn More

Dextrose, Monohydrate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5391173
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002700 500 g Poly Bottle
02002699 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002700 Supplier Avantor Supplier No. 191001
Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 1496309
PubChem CID 133126654

Specifications

Packaging Poly Bottle
Molecular Formula C6H14O7
Quantity 500 g
Specific Gravity 1L = 1.54kg
Synonym 6-hydroxymethyl oxane-2,3,4,5-tetrol hydrate, d-galactose hydrate, dextrose monohydrate, usp, .alpha.-d-glucose hydrate, dextrose, u.s.p.
InChI Key OSNSWKAZFASRNG-WNFIKIDCSA-N
SMILES C(C1C(C(C(C(O1)O)O)O)O)O.O
IUPAC Name (2R,3S,4R,5R,6S)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol;hydrate
Molecular Weight (g/mol) 198.17
Formula Weight 198.17
Grade BAKER ANALYZED™ Reagent
Physical Form Powder
Chemical Name or Material Dextrose, Monohydrate
Show More Show Less