missing translation for 'onlineSavingsMsg'
Learn More

Dextrose (D-Glucose), Anhydrous (Granular Powder/Certified ACS), Fisher Chemical™

Catalog No. p-98685
Change view
Click to view available options
Quantity:
500 g
1 kg
3 kg
10 kg
50 kg
Packaging:
Fiber Drum
Glass Bottle
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D16500 500 g Glass Bottle
D161 1 kg Poly Bottle
D163 3 kg Poly Bottle
D1610 10 kg Poly Pail
D1650 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. D16500 Supplier Thermo Fisher Scientific Supplier No. D16 500
Edge

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00148912
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 146.1°C
Color White
pH 5.9
Ignition Residue 0.02% max.
Loss on Drying 0.2% max.
Packaging Glass Bottle
Linear Formula CH2OH(CHOH)4CHO
Quantity 500 g
Specific Rotation +52.5 to +53.0° (+ or −)
Solubility Information 0.005% max. Insoluble Matter
Vapor Pressure Negligible
Formula Weight 180.16
Grade Certified ACS
Physical Form Solid
Chemical Name or Material Dextrose (D-Glucose) Anhydrous
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0