Learn More
Dextrose (D-Glucose), Anhydrous (Granular Powder/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific D16 50
| Quantity | 50 kg |
|---|---|
| Packaging | Fiber Drum |
Chemical Identifiers
| C6H12O6 | |
| MFCD00148912 | |
| 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
| CHEBI:42758 | |
| C(C(C(C(C(C=O)O)O)O)O)O |
Specifications
| Dextrose (D-Glucose) Anhydrous | |
| 146.1°C | |
| 5.9 | |
| 0.2% max. | |
| C6H12O6 | |
| 50 kg | |
| +52.5 to +53.0° (+ or −) | |
| 0.005% max. Insoluble Matter | |
| C(C(C(C(C(C=O)O)O)O)O)O | |
| 180.156 | |
| CHEBI:42758 | |
| 180.16 | |
| Solid |
| 50-99-7 | |
| White | |
| 0.02% max. | |
| Fiber Drum | |
| CH2OH(CHOH)4CHO | |
| MFCD00148912 | |
| 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
| GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
| (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal | |
| 107526 | |
| Negligible | |
| Certified ACS |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0