missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrose (D-Glucose), Anhydrous (Granular Powder/Certified ACS), Fisher Chemical™
Specifications
Specifications
| Melting Point | 146.1°C |
| Color | White |
| pH | 5.9 |
| Ignition Residue | 0.02% max. |
| Loss on Drying | 0.2% max. |
| Packaging | Poly Bottle |
| Linear Formula | CH2OH(CHOH)4CHO |
| Quantity | 3 kg |
| Specific Rotation | +52.5 to +53.0° (+ or −) |
| Solubility Information | 0.005% max. Insoluble Matter |
| Show More |
Safety and Handling
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. . NFPA Health:1 Flammability:0 Instability:0 |