missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrose, Anhydrous, Ultrapure, 97.5-102.0%, Spectrum™ Chemical
$809.89 - $2,043.79
Chemical Identifiers
| CAS | 50-99-7 |
|---|---|
| Molecular Formula | C6H12O6 |
| Molecular Weight (g/mol) | 180.16 |
| InChI Key | WQZGKKKJIJFFOK-UHFFFAOYNA-N |
| IUPAC Name | 6-(hydroxymethyl)oxane-2,3,4,5-tetrol |
| SMILES | OCC1OC(O)C(O)C(O)C1O |
Chemical Identifiers
| 50-99-7 | |
| 180.16 | |
| 6-(hydroxymethyl)oxane-2,3,4,5-tetrol |
| C6H12O6 | |
| WQZGKKKJIJFFOK-UHFFFAOYNA-N | |
| OCC1OC(O)C(O)C(O)C1O |
Specifications
| 50-99-7 | |
| 0.001 | |
| WQZGKKKJIJFFOK-UHFFFAOYNA-N | |
| 6-(hydroxymethyl)oxane-2,3,4,5-tetrol | |
| 97.5 to 102.0% | |
| To Pass Test |
| 1 | |
| C6H12O6 | |
| OCC1OC(O)C(O)C(O)C1O | |
| 180.16 | |
| Ultrapure |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.