missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrose, Anhydrous, U.S.P., J.T. Baker™
Specifications
Specifications
| Packaging | Poly Bottle |
| Quantity | 500 g |
| Specific Gravity | 1L = 1.54kg |
| Formula Weight | 180.16 |
| Percent Purity | 97.5 to 102.0% |
| Grade | USP |
| Physical Form | Solid |
| Chemical Name or Material | Dextrose, Anhydrous |