missing translation for 'onlineSavingsMsg'
Learn More

Dextrose, Anhydrous, Multi-Compendial, U.S.P., J.T. Baker™

Catalog No. 02002710
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02002710 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02002710 Supplier Avantor Supplier No. 192005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

GMP-manufactured dextrose meeting USP specifications.

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Packaging Poly Bottle
Quantity 2.5 kg
Specific Gravity 1L = 1.54kg
Formula Weight 180.16
Percent Purity 97.5 to 102.0%
Grade Multi-Compendial/USP
Physical Form Solid
Chemical Name or Material Dextrose, Anhydrous