missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrose, Anhydrous, Granular, BiotechGrade, Spectrum™ Chemical
D1082, 50-99-7, C6H12O6
$435.34 - $5845.71
Chemical Identifiers
| CAS | 50-99-7 |
|---|---|
| Molecular Formula | C6H12O6 |
| Molecular Weight (g/mol) | 180.16 |
| InChI Key | WQZGKKKJIJFFOK-UHFFFAOYNA-N |
| IUPAC Name | 6-(hydroxymethyl)oxane-2,3,4,5-tetrol |
| SMILES | OCC1OC(O)C(O)C(O)C1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602109
|
Spectrum Chemical Mfg Cor
D1082500GM |
500 g | Poly Bottle |
Each for $435.34
|
|
||||
|
18602110
|
Spectrum Chemical Mfg Cor
D108225KG |
2.5 kg | Poly Bottle |
Each for $1,253.49
|
|
||||
|
18602111
|
Spectrum Chemical Mfg Cor
D108212KG |
12 kg | Poly Pail |
Each for $3,383.20
|
|
||||
|
18602112
|
Spectrum Chemical Mfg Cor
D108245KGBL |
45 kg | Fiber Drum |
Each for $5,845.71
|
|
||||