missing translation for 'onlineSavingsMsg'
Learn More

Dextrose Anhydrous (Crystalline Granules/Molecular Biology), Fisher BioReagents

Catalog No. p-25474
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP350500 500 g Glass Bottle
BP3501 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP350500 Supplier Fisher BioReagents Supplier No. FLBP350500
Only null left
Dextrose is often used as a cell culture additive.

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 146.1°C
Color White
pH 5.9
Assay Percent Range ≥99.8 %
Packaging Glass Bottle
Linear Formula HOCH2(CHOH)4CHO
Quantity 500 g
Merck Index 15, 4495
Specific Rotation 52.5 to 53.0° (+ or -)
Solubility Information 0.005% max. Insoluble Matter
Vapor Pressure Negligible
Formula Weight 180.08
Percent Purity ≥99.8%
Grade DNase-, RNase- and Protease-Free
Physical Form Solid
Chemical Name or Material Dextrose Anhydrous
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
Recommended Storage RT