missing translation for 'onlineSavingsMsg'
Learn More

Dextrin, White Powder, Spectrum™ Chemical

Catalog No. 18602098
Change view
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602098 500 g Poly Bottle
18602099 2.5 kg Poly Bottle
18602100 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18602098 Supplier Spectrum Chemical Mfg Cor Supplier No. DE137500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

DE137, 9004-53-9, (C6H10O5)n·xH2O

Spectrum™ Chemical Dextrin, White Powder , is used as water-soluble glue in envelope adhesives, crispness enhancers in food processing and as a thickening agent in pharmaceuticals. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9004-53-9
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.44
InChI Key FYGDTMLNYKFZSV-MRCIVHHJSA-N
IUPAC Name (3R,4S,5S,6R)-2-{[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1OC(O[C@H]2[C@H](O)[C@@H](O)C(O[C@H]3[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

CAS Max % 100%
Quantity 500 g
Grade Reagent
Packaging Poly Bottle