accessibility menu, dialog, popup

UserName

HO(R)OO(S)(R)OH(R)OHO(R)HOO(S)(R)OH(R)OH(S)HOO(R)OH(R)OH(S)OH(S)OH

CAS RN 9004-53-9

IUPAC Name: (3R,4S,5S,6R)-2-{[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: microcrystalline cellulose emdex 30an45 dextrin 101 dextrina bianca farinex CO 2 fungal amylase starch corn dextrin arabix 6 arabix 7 cream dextrin 15
Molecular Weight (g/mol): 504.44
Molecular Formula: C18H32O16
InChi Key: FYGDTMLNYKFZSV-MRCIVHHJSA-N
SMILES: OC[C@H]1OC(O[C@H]2[C@H](O)[C@@H](O)C(O[C@H]3[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Grade 
  • (6)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (6)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (4)
  • (5)
  • (3)
  • (6)
  • (1)