missing translation for 'onlineSavingsMsg'
Learn More

Dextrin, precipitated by alcohol

Catalog No. p-7022802
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1571722 100 g
AAA1571736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1571722 Supplier Thermo Scientific Chemicals Supplier No. A1571722
Only null left

CAS: 9004-53-9 | C18H32O16 | 504.438 g/mol

Dextrin is used as adhesive in the manufacture of gummed tapes, textiles and paper. It is used in producing nutritional products. It is also used as a absorbents, binding agents and viscosity controlling agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-53-9
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
MDL Number MFCD00081554
InChI Key FYGDTMLNYKFZSV-MRCIVHHJSA-N
Synonym dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen
PubChem CID 62698
IUPAC Name (3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O

Specifications

Merck Index 14,2953
Solubility Information Soluble in hot water (0.1g/ml).
Quantity 100 g
Sensitivity Hygroscopic
Chemical Name or Material Dextrin, precipitated by alcohol
EINECSNumber 232-675-4
RTECSNumber HH9450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only