missing translation for 'onlineSavingsMsg'
Learn More

Dextran, for biochemistry, low fraction

Catalog No. AC406271000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406271000 100 g Plastic bottle
AC406275000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC406271000 Supplier Thermo Scientific Chemicals Supplier No. 406271000

CAS: 9004-54-0 | (C6H9O5)n(C6H10O5)m | 504.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-54-0
Molecular Formula (C6H9O5)n(C6H10O5)m
Molecular Weight (g/mol) 504.44
MDL Number MFCD00130935
InChI Key FZWBNHMXJMCXLU-UHFFFAOYNA-N
Synonym dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
PubChem CID 4125253
IUPAC Name Dextran
SMILES O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O

Specifications

Color White
pH 4.5 to 7.0 (6% aq. soln.)
Physical Form Powder
Ash 2.0% max.
Avg. Mol. Wt. or Mol. Wt. Range 60000 to 90000
Linear Formula C6H10O5
Quantity 100 g
Grade Biochemical
Infrared Spectrum Authentic
Loss on Drying 7.0% max.
Packaging Plastic bottle
Viscosity 0.22 to 0.27 (1% sol, 37°C)
Chemical Name or Material Dextran, Low fraction, For biochemistry
Show More Show Less

RUO – Research Use Only