missing translation for 'onlineSavingsMsg'
Learn More

Dextran Sulfate Sodium Salt

CAS: 9011-18-1 | (C6H7O5)m(C6H7O5)n | 40,000 g/mol

Supplier:  Thermo Fisher Scientific J1448918

 View more versions of this product

Catalog No. AAJ1448918


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dextran Sulfate Sodium Salt
Powder
(C6H7O5)m(C6H7O5)n
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
40,000
CHEBI:116735
Plastic bottle
9011-18-1
50 g
MFCD00081551
BLFLLBZGZJTVJG-UHFFFAOYSA-N
ethyl 4-aminobenzoate
2337
40,000- 50,000

Chemical Identifiers

9011-18-1
40,000
BLFLLBZGZJTVJG-UHFFFAOYSA-N
2337
ethyl 4-aminobenzoate
(C6H7O5)m(C6H7O5)n
MFCD00081551
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
CHEBI:116735
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : HH9290000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only