missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran Sulfate Sodium Salt
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 100 g |
| Packaging | Plastic bottle |
| Formula Weight | 40,000- 50,000 Kd |
| Grade | Ungraded |
| Chemical Name or Material | Dextran Sulfate Sodium Salt |
Safety and Handling
| RTECSNumber | HH9290000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only