missing translation for 'onlineSavingsMsg'
Learn More

Dextran Sulfate Sodium Salt

Catalog No. AAJ1448909
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ1448909 10 g Plastic bottle
AAJ1448918 50 g Plastic bottle
AAJ1448922 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ1448909 Supplier Thermo Fisher Scientific Supplier No. J1448909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9011-18-1 | (C6H7O5)m(C6H7O5)n | 40,000 g/mol

Chemical Identifiers

CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 40,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
IUPAC Name ethyl 4-aminobenzoate
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

Physical Form Powder
Quantity 10 g
Formula Weight 40,000- 50,000
Packaging Plastic bottle
Chemical Name or Material Dextran Sulfate Sodium Salt
RTECSNumber HH9290000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only