Learn More
Dextran Sulfate Sodium Salt, Powder, Spectrum™ Chemical
DE133, 9011-18-1
Supplier:  Spectrum Chemical Mfg Cor DE13325GM
 View more versions of this product
Catalog No. 18602077
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Dextran Sulfate Sodium Salt, Powder is a sodium salt of the sulfuric acid ester of Dextran with an average molecular weight of 12,000. It functions as a nonsurfactant, suspending agent with gel forming and viscosity controlling properties. It can be used in gels and moisturizers. It appears as a white micro-crystalline powder that is soluble in cold water.
Specifications
Specifications
9011-18-1
(C6H7O5)m(C6H7O5)n
25 g
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
40,000
Poly Bottle
100%
MFCD00081551
BLFLLBZGZJTVJG-UHFFFAOYSA-N
Dextran sulfate sodium salt
Reagent
Chemical Identifiers
9011-18-1
40,000
BLFLLBZGZJTVJG-UHFFFAOYSA-N
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
(C6H7O5)m(C6H7O5)n
MFCD00081551
Dextran sulfate sodium salt
Product Suggestions
Product Suggestions