missing translation for 'onlineSavingsMsg'
Learn More

Dextran Sulfate Sodium Salt, Powder, Spectrum™ Chemical

Catalog No. 18602076
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602076 5 g Amber Glass Bottle
18602077 25 g Poly Bottle
18602078 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18602076 Supplier Spectrum Chemical Mfg Cor Supplier No. DE1335GM
Only null left

DE133, 9011-18-1

Spectrum™ Chemical Dextran Sulfate Sodium Salt, Powder is a sodium salt of the sulfuric acid ester of Dextran with an average molecular weight of 12,000. It functions as a nonsurfactant, suspending agent with gel forming and viscosity controlling properties. It can be used in gels and moisturizers. It appears as a white micro-crystalline powder that is soluble in cold water.

Chemical Identifiers

CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 40,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
IUPAC Name Dextran sulfate sodium salt
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

CAS Max % 100%
Quantity 5 g
Grade Reagent
Packaging Amber Glass Bottle