missing translation for 'onlineSavingsMsg'
Learn More

Dextran sulfate sodium salt, MW ca 8,000

CAS: 9011-18-1 | (C6H7O5)m(C6H7O5)n

Supplier:  Thermo Scientific Chemicals J6210122

 View more versions of this product

Catalog No. AAJ6210122


Only null left
Explore available promotions

Description

Description

Dextran sulfate is used asa nucleic acid hybridization accelerant. The ability of the polymer to precipitate fibrinogen and low-density lipoproteins from plasma solutions has been used for analysis and for anticoagulant applications. Dextran Sulfate Sodium (DSS) which has proven to have versatile value in Biomedical/clinical research. DSS can be used to study Colitis (Inflammatory Bowel disease) as well as an antiviral as agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

White to Yellow
Powder
(C6H7O5)m(C6H7O5)n
14,2951
Soluble in water (100mg/ml).
BLFLLBZGZJTVJG-UHFFFAOYSA-N
Dextran sulfate sodium salt
CHEBI:116735
Phenol-like
Dextran sulfate sodium salt
9011-18-1
MFCD00081551
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
100 g
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
2337
≈8 kd

Chemical Identifiers

9011-18-1
MFCD00081551
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
CHEBI:116735
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
(C6H7O5)m(C6H7O5)n
BLFLLBZGZJTVJG-UHFFFAOYSA-N
2337
Dextran sulfate sodium salt
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : HH9290000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only